missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Petroleum ether, Laboratory Reagent, bp 40-60 °C, Honeywell™
Click to view available options
Quantity:
5 L
4 x 5 L
Packaging:
Aluminum bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Color | Colorless |
Density | 0.65g/cu.cm at 20°C |
Boiling Point | 40°C to 60°C (1013 hPa) |
Flash Point | −30°C (−22°F) |
Odor | Characteristic |
Packaging | Aluminum bottle |
Quantity | 4 x 5 L |
UN Number | UN1268 |
Synonym | isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane |
Solubility Information | Practically insoluble in water |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'