missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGM3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522353
This item is not returnable.
View return policy
Description
Recommended positive controls: NT2D1, IMR32, Jurkat, HeLa, mouse brain. Predicted reactivity: Mouse (84%), Rat (81%), Chimpanzee (100%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Catalyzes the conversion of GlcNAc-6-P into GlcNAc-1-P during the synthesis of uridine diphosphate/UDP-GlcNAc, a sugar nucleotide critical to multiple glycosylation pathways including protein N- and O-glycosylation.Specifications
PGM3 | |
Polyclonal | |
Unconjugated | |
Pgm3 | |
2810473H05Rik; acetylglucosamine phosphomutase; Agm1; BB187688; C77933; GlcNAc-P mutase; IMD23; N-acetylglucosamine-phosphate mutase; N-acetylglucosamine-phosphate mutase 1; PAGM; PGM 3; Pgm3; Pgm-3; Phosphoacetylglucosamine mutase; phosphoglucomutase 3; phosphoglucomutase-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
109785, 5238 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.73 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O95394, Q9CYR6 | |
Pgm3 | |
Recombinant fragment corresponding to a region within amino acids 44 and 321 of PGM3 (Uniprot ID#O95394) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |