missing translation for 'onlineSavingsMsg'
Learn More
Buffer Solution, pH 4.00, Chem-Lab Product Code.: 10335792

Buffer Solution, pH 4.00, Chem-Lab

Product Code. 10335792
1000 mL, 1 L
Click to view available options
Quantity:
1000 mL
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 10335792

Brand: Chem Lab CL03.0213.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for pH measurement and maintenance in moderately acidic samples. Buffer Solution, pH 4.00, Chem-Lab is a solution of citrate, sodium chloride and sodium hydroxide, with a pH level of 4.00 ±0.02 at 20°C. Solution is traceable to standard reference materials (SRM) from NIST.

pH Levels ± 0.02:
  • pH 4.03 — 0°C
  • pH 4.02 — 10°C
  • pH 4.00 — 20°C
  • pH 4.00 — 25°C
  • pH 4.00 — 30°C
  • pH 4.00 — 40°C
  • pH 4.00 — 50°C
  • pH 4.00 — 60°C
  • pH 4.00 — 70°C
  • pH 4.00 — 80°C
  • pH 4.00 — 90°C

Specifications

Melting Point 0°C
Boiling Point 100°C
Color Colorless
Density 1.01g/mL
For Use With (Application) Laboratory use, pH measurements
pH 4.00 ±0.02
Physical Form Liquid
Quantity 1000 mL
Solution Type pH Buffer
Formulation Citrate-NaCl-NaOH
Odor Odorless
Packaging Polyethylene
Traceability to NIST Yes
Solubility Information Soluble in water
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer Solution, pH 4.00, Chem-Lab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.