Learn More
Mettler Toledo™ pH Combination Electrodes
Brand:  Mettler Toledo™ 59903230
4117.71 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10425072
  • 10535.00 DKK / Each
Only null left
Estimated Shipment: 29-07-2025
Add to basket
This item is not returnable. View return policy
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
pH Combination Electrode
0 to 12
Use in reactors, tanks, open basins and transfer lines under harsh chemical process conditions.
120 mm
0°C to 100°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mettler Toledo™ pH Combination Electrodes > Length: 120mm, pH Range: 0 to 12