missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ L/S™ PharMed™ BPT Spooled Pump Tubing

Use this spooled tubing for cell and tissue culture work and pharmaceutical uses. Masterflex™ L/S™ PharMed™ BPT Spooled Pump Tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport.

Brand:  Masterflex™ 95687-18

 View more versions of this product

Product Code. 11735248

  • 22655.00 DKK / Each
Estimated Shipment: 02-09-2025
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Opaque beige spooled pump tubing requires less storage space than the equivalent amount of individual tubing coils
  • Heat-sealable, bondable and formable
  • Ideal for light-sensitive samples
  • Nontoxic, nonhemolytic, extremely low permeability
  • Resists acids, alkalis, oxidizing agents and animal and vegetable oils
  • Reduce scrap, cut only as much as you need
  • Ensure that you always have enough tubing on hand
Nearly any Masterflex pump tubing formulation can be spooled. Call our Application Specialists for a quote or to place a special order
Specifications

Specifications

FDA approved; Prop 65 NA (no Prop 65 chemicals)
7.9 mm
121.9 m
18
3/8 in.
Beige
510 mm Hg (20 in. Hg)
Pump Tubing
Autoclavable
400 ft.
-20°C
2300 mL/min
Autoclave, Ethylene Oxide, Gamma Irradiation
15 psi (1 bar)
100°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Masterflex™ L/S™ PharMed™ BPT Spooled Pump Tubing > 18

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.