missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Phenol Test Kits
Brand: WTW™ 250412
999.77 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
4-Amoniantupyrin | |
PhotoLab™ S12, PhotoLab™ Spectral photometers | |
25 | |
Photometric |
For waste water, surface water | |
Certificates | |
25 Tests | |
CombiCheck Phenol (C6H5OH) cell test kit |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Phenol Cell Test Method: photometric 0.10 - 2.50 mg/l Spectroquant
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTW™ Phenol Test Kits > Cell Test; Detection Range: 0.10 to 2.5mg/L
Spot an opportunity for improvement?Share a Content Correction