Learn More
Phenyltrimethylammonium iodide, 99%, Thermo Scientific Chemicals Product Code.: 11447017
Phenyltrimethylammonium iodide, 99%, Thermo Scientific Chemicals
Product Code. 11447017
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
Product Code. 11447017
Brand: Thermo Scientific Alfa Aesar A14402.22
Request Bulk or Custom Format
Phenyltrimethylammonium iodide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenyltrimethylammonium iodide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.
Solubility
Soluble in water and methanol.
Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. It is hygroscopic in nature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 98-04-4
Molecular Formula C9H14IN
Molecular Weight (g/mol) 263.122
MDL Number MFCD00011791
InChI Key KKLAORVGAKUOPZ-UHFFFAOYSA-M
Synonym phenyltrimethylammonium iodide, trimethylphenylammonium iodide, n,n,n-trimethylanilinium iodide, trimethylanilinium iodide, trimethylanillinium iodide, n,n-dimethylaniline methiodide, unii-4o888xq7fp, ammonium, phenyltrimethyl-, iodide, n,n,n-trimethylbenzenaminium iodide, benzenaminium, n,n,n-trimethyl-, iodideShow MoreShow Less
PubChem CID 94135
IUPAC Name trimethyl(phenyl)azanium;iodide
SMILES C[N+](C)(C)C1=CC=CC=C1.[I-]
Specifications
Melting Point ∼227°C (sublimation)
Quantity 100 g
UN Number UN2811
Beilstein 3916752
Sensitivity Light sensitive
Merck Index 14,7318
Solubility Information Soluble in water and methanol.
Formula Weight 263.12
Percent Purity 99%
Chemical Name or Material Phenyltrimethylammonium iodide
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10247697 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog NumberA14402.22,A14402.14,A14402.36
Lot Number10243361 Certificate TypeCertificate of Analysis Date12/09/2022 Catalog NumberA14402.22,A14402.14,A14402.36
Lot Number10240087 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14402.22,A14402.14,A14402.36
Lot Number10232914 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14402.22,A14402.14,A14402.36
Lot NumberL02X023 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14402.22,A14402.14,A14402.36
compliance-icons
Product Identifier
  • Phenyltrimethylammonium iodide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Phenyltrimethylammonium iodide
RUO – Research Use Only
Product Content Correction
Product Title
Phenyltrimethylammonium iodide, 99%, Thermo Scientific Chemicals >