missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Phosphate, 0.5M buffer soln., pH 7.2
Click to view available options
Quantity:
250 mL
500 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Concentration | 0.5M buffer soln. |
Molecular Weight (g/mol) | 119.976 |
Quantity | 250 mL |
Packaging | Plastic Bottle |
Content And Storage | Ambient Temperatures |
Form | Liquid |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals Phosphate, 0.5M buffer soln., pH 7.2 >
Spot an opportunity for improvement?Share a Content Correction