missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Phosphate, 0.5M buffer soln., pH 9.0
483.64 DKK - 651.60 DKK
Specifications
Concentration | 0.5M buffer soln. |
---|---|
Molecular Weight (g/mol) | 119.976 |
Packaging | Plastic Bottle |
CAS | 7558-80-7 |
Content And Storage | Ambient Temperatures |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15424749
|
Thermo Scientific™
J62345.AK |
250 mL |
483.64 DKK
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15434749
|
Thermo Scientific™
J62345.AP |
500 mL |
651.60 DKK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
0.5M buffer soln. | |
Plastic Bottle | |
Ambient Temperatures |
119.976 | |
7558-80-7 | |
Liquid |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Phosphate, 0.5M buffer soln., pH 9.0