Learn More
Medicago Phosphate Buffered Saline Tablets with Tween™ 20, pH 7.4
Supplied as exactly pre-weighed tablets with a final buffer volume of 500 or 1000ml. Medicago Phosphate Buffered Saline Tablets with Tween™ 20, pH 7.4 is dissolved in deionized water to yield a solution of sodium chloride, potassium chloride, Tween™ 20 and phosphate buffer.
2625.00 DKK - 3940.00 DKK
Specifications
Form | Tablets |
---|---|
For Use With (Application) | ELISA, Western Blot, Plate-based Assays, Protein-plate Coating |
Formulation | NaCl (0.14M), KCl (0.0027M), PO4 (0.01M), Tween™ 20 (0.05%) |
pH Range | 7.4 at 25°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11300439
|
Medicago
09-9410-100 |
100 x 1000 mL Tablets |
2625.00 DKK
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10602203
|
Medicago
09-9410-100 |
100 x 1000 mL Tablets |
3940.00 DKK
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PBS-T buffer contains the non-ionic detergent Tween™ 20 which acts through blocking. It has the ability to reduce non-specific binding and protein-protein interaction during the wash step in protein and immunoassay procedures such as ELISA and Western blotting. Decreasing the non-specific binding and staining makes ELISA results and blots easier to interpret.
- Formulated from analytical grade reagents
- PBS with non-ionic detergent
- Specifically developed for immunoassays
- Exactly pre-weighed tablets
- Dissolve and go for greater convenience
Specifications
Tablets | |
ELISA, Western Blot, Plate-based Assays, Protein-plate Coating | |
NaCl (0.14M), KCl (0.0027M), PO4 (0.01M), Tween™ 20 (0.05%) | |
7.4 at 25°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.