Learn More
WTW™ Phosphate Test Kits
Obtain precision measurements for waste water, drinking water, surface water, and fertilizers with CombiCheck Phosphate (PO4) cell test kits.
Brand:  WTW™ 250446
1238.52 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10327481
  • 1275.00 DKK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Variety of instrumentation for measuring phosphates
  • PhotoLab® S6, PhotoLab® S12, PhotoLab® Spectral, Photoflex photometers
  • For waste water, drinking water, surface water, fertilizers
  • CombiCheck Phosphate (PO<sub>4</sub>) cell test kit
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Chromosulfuric acid oxidation
For PhotoLab™ S12, PhotoLab™ Spectral, spectroFlex, Photoflex photometers
95
25 Tests
CombiCheck COD Chemical oxygen demand O2 cell test kit
For waste water, drinking water, surface water, fertilizers
Certificates
0.010 to 5 mg/L PO4-P, 0.03 to 15.3 mg/L PO4
Photometric
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTW™ Phosphate Test Kits > Reagent Test; Detection Range: 0.010 to 5mg/L PO4-P, 0.03 to 15.3mg/L POsub4