Learn More
WTW™ Phosphate Test Kits
Obtain precision measurements for waste water, drinking water, surface water, and fertilizers with CombiCheck Phosphate (PO4) cell test kits.
Brand: WTW™ 250413
1014.69 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
View more versions of this product
Description
- Variety of instrumentation for measuring phosphates
- PhotoLab® S6, PhotoLab® S12, PhotoLab® Spectral, Photoflex photometers
- For waste water, drinking water, surface water, fertilizers
- CombiCheck Phosphate (PO<sub>4</sub>) cell test kit

Specifications
Vanadate Molybdate | |
PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers | |
25 | |
25 Tests | |
CombiCheck Phosphate (PO4) cell test kit |
For waste water, drinking water, surface water, fertilizers | |
Certificates | |
0.5 to 25 mg/L PO4-P, 1.5 to 76.7 mg/L PO4 | |
Photometric |
Safety and Handling
- Phosphate Cell Test (o-phosphate)
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid, Ammonium monovanadate
Your input is important to us. Please complete this form to provide feedback related to the content on this product.