missing translation for 'onlineSavingsMsg'
Learn More

Phospho-EIF4B (Ser406) Rabbit Recombinant Antibody, Proteintech

Rabbit Recombinant Antibody

968.42 DKK - 2995.23 DKK

Specifications

Antigen EIF4B (Ser406)
Clone 5G14
Concentration 300 ÎĽg/mL
Applications Western Blot, Immunohistochemistry, ELISA
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30237247
View Documents
Proteintech
80059-1-RR-100UL
100 ÎĽL
2995.23 DKK
100µL
Please to purchase this item. Need a web account? Register with us today!
30236459
View Documents
Proteintech
80059-1-RR-20UL
20 ÎĽL
968.42 DKK
20µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Phospho-EIF4B (Ser406) antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human Phospho-EIF4B (Ser406). This antibody recognizes human, mouse, rat antigen. The Phospho-EIF4B (Ser406) antibody has been validated for the following applications: WB, IHC, ELISA analysis.
Specifications

Specifications

EIF4B (Ser406)
300 ÎĽg/mL
Recombinant Monoclonal
Liquid
RUO
1975
Peptide
Primary
Store at -20°C. Stable for one year after shipment.
5G14
Western Blot, Immunohistochemistry, ELISA
Unconjugated
Rabbit
Human, Mouse, Rat
EIF-4B; PRO1843
IgG
Protein A purification
Recombinant Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phospho-EIF4B (Ser406) Rabbit Recombinant Antibody, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.