missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Phosphopantothenate-cysteine ligase Protein
Highly purified. Generating reliable and reproducible results.
Brand: Novus Biologicals™ NBP1-78806-100UG
This item is not returnable.
View return policy
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-311 of Human Phosphopantothenate-cysteine ligase The Recombinant Human Phosphopantothenate-cysteine ligase Protein is derived from E. coli. The Recombinant Human Phosphopantothenate-cysteine ligase Protein has been validated for the following applications: SDS-Page.Specifications
1.0mg/mL | |
Liquid. 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol | |
SDS-PAGE | |
Human | |
RUO | |
Unconjugated |
SDS-PAGE | |
36.1kDa | |
100μg | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Phosphopantothenate-cysteine ligase | |
>95% |