missing translation for 'onlineSavingsMsg'
Learn More
Phthalic anhydride, ACS reagent Product Code.: 10188130

Phthalic anhydride, ACS reagent

Product Code. 10188130 Shop All Thermo Scientific Chemicals Products
2 kg, Glass bottle
Click to view available options
Quantity:
500 g
2 kg
5 kg
Packaging:
Glass bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10188130

Brand: Thermo Scientific Acros 423320020

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85-44-9 | C8H4O3 | 148.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-44-9
Molecular Formula C8H4O3
Molecular Weight (g/mol) 148.12
MDL Number MFCD00005918
InChI Key LGRFSURHDFAFJT-UHFFFAOYSA-N
Synonym phthalic anhydride, isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 1,3-dioxophthalan, 1,3-phthalandione, phthalsaeureanhydrid, phthalic acid anhydride, o-phthalic acid anhydride, phthalandione, retarder esen
PubChem CID 6811
ChEBI CHEBI:36605
SMILES O=C1OC(=O)C2=CC=CC=C12

Specifications

Melting Point 129.0°C to 132.0°C
Color White
Boiling Point 284.0°C
Flash Point 152°C
Ignition Residue 0.01% max.
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 2 kg
Beilstein 17,469
Fieser 01,882; 17,148
Merck Index 15,7484
Solubility Information Solubility in water: 6g/L (20°C). Other solubilities: soluble in alcohol and carbon disulfide, slightly soluble in ether, 7.5% in benzene (25°C), forms phthalic acid in water
IUPAC Name 2-benzofuran-1,3-dione
Viscosity 6.4 mPa.s (197°C)
Formula Weight 148.12
Percent Purity 99.0 to 100.2%
Grade ACS Reagent
Physical Form Flaky Crystals
Chemical Name or Material Phthalic anhydride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0457682 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog Number423325000,423320050,423320020
Lot NumberA0465221 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number423325000,423320050,423320020
Lot NumberA0457682 Certificate TypeCertificate of Analysis Date09/11/2023 Catalog Number423325000,423320050,423320020
Lot NumberA0457681 Certificate TypeCertificate of Analysis Date09/11/2023 Catalog Number423325000,423320050,423320020
Lot NumberA0448762 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number423325000,423320050,423320020
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phthalic anhydride, ACS reagent >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.