Learn More
uvex Phynomic XG Safety Gloves with Aqua-Polymer Xtra Grip Foam Coating
Shop All Uvex Products
Click to view available options
Size:
6
7
8
9
10
11
12
Description
- Xtra Grip aqua-polymer foam coating is breathable and provides a high level of mechanical abrasion resistance
- Enhanced grip capabilities in oily areas
- Tactile feel when assembling oily parts
- Ideal for precision work, maintenance, repair work, and metal processing
Greener Choice Claims
- Environmental benefits include:
- Recycled Content
- Free-of a substance that causes environmental harm
- Renewable Energy
Specifications
Specifications
Construction | Seamless Knit |
Coating Material | Aqua-Polymer Foam |
Color | Black, Gray |
Exterior Finish | Smooth |
Hand | Ambidextrous |
Latex Free | Yes |
Material | Spandex, Polyamide |
Product Type | Mechanical Protection Gloves |
Size | 7 |
Style | Full-hand Mitt |
Show More |
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
uvex Phynomic XG Safety Gloves with Aqua-Polymer Xtra Grip Foam Coating >
Spot an opportunity for improvement?Share a Content Correction