Learn More
Thermo Scientific™ Pierce™ Alkaline Phosphatase, Biotinylated
Purified biotin and biotinylated BSA, HRP, AP or FITC for use as controls or to amplify signal in IHC via the avidin-biotin complex (ABC).
Brand:  Thermo Scientific™ 29339
Product Code. 10148103
  • 2520.00 DKK / 1mg
Explore more special offers
Description
Description
Thermo Scientific™ Pierce Biotinylated Proteins include biotin-labeled horseradish peroxidase (B-HRP), alkaline phosphatase (B-AP) and fluorescein (B-FITC) for use as controls or signal amplification in IHC via avidin-biotin complex (ABC) techniques.
Biotinylated HRP and biotinylated AP are most commonly used in immunohistochemistry (IHC) to amplify the signal of biotinylated primary antibodies using the ABC staining method. Biotinylated Fluorescein (FITC) cannot be used to polymerize and amplify the signal to the same degree as biotinylated enzymes because each fluor molecule is tagged with only one biotin molecule. However, the fluorescent variant of traditional ABC staining is possible with this unique molecule.
Highlights:
  • Biotinylated enzymes – biotin-labeled horseradish peroxidase (B-HRP), alkaline phosphatase (B-AP) and beta-galactosidase (B-bGal) for use in avidin-biotin complex (ABC) staining
  • Biotinylated FITC – amplify the fluorescein signal of biotinylated primary antibodies by using avidin or streptavidin plus biotinylated fluorescein (B-FITC)
  • Biotinylated controls – use biotinylated bovine serum albumin (B-BSA) or any one of these molecules as positive controls in biotin quantitation methods
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at or below -20°C.
Pierce Alkaline Phosphatase, Biotinylated
1 mg
Pierce™
Biotin & Analogs
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZG391862 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog Number29339
Lot NumberYL380903 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number29339
Lot NumberYE372944 Certificate TypeCertificate of Analysis Date23/06/2023 Catalog Number29339
Lot NumberXH361235 Certificate TypeCertificate of Analysis Date27/09/2022 Catalog Number29339
Lot NumberWJ333875 Certificate TypeCertificate of Analysis Date01/06/2022 Catalog Number29339
Product Title
Thermo Scientific™ Pierce™ Alkaline Phosphatase, Biotinylated > Alkaline Phosphatase, Biotinylated; 1mg