missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GMBS (N-Îł-maleimidobutyryl-oxysuccinimide ester)

Product Code. 10772915 Shop All Thermo Scientific Products
50 mg, 50mg
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Crosslink amines to sulfhydryls with this short crosslinker composed of NHS-ester and maleimide groups at ends of an aliphatic, 6-atom (7.3A) spacer arm.

Thermo Scientific™ Pierce GMBS is an amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a short spacer arm (7.3 Ås).

Highlights:

  • Less immunogenic than SMCC
  • Reactive groups: NHS ester and maleimide
  • Reactive toward: amino and sulfhydryl groups
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Sulfhydryl
Cleavable No
Description GMBS
Molecular Weight (g/mol) 280.23
PEGylated No
Spacer Arm Length 7.3 AĚŠ
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, NHS Ester
Spacer Short (<10 AĚŠ)
Form Powder
Quantity 50 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less

Manuals

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberZK407016 Certificate TypeCertificate of Analysis Date16/12/2024 Product Code22309
Lot NumberZJ372162 Certificate TypeCertificate of Analysis Date05/11/2024 Product Code22309
Lot NumberYK380264 Certificate TypeCertificate of Analysis Date07/12/2023 Product Code22309
Lot NumberYF375605 Certificate TypeCertificate of Analysis Date21/07/2023 Product Code22309
Lot NumberYD369948 Certificate TypeCertificate of Analysis Date03/05/2023 Product Code22309

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ GMBS (N-Îł-maleimidobutyryl-oxysuccinimide ester) >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.