Learn More
Thermo Scientific™ Pierce™ High Sensitivity NeutrAvidin™-HRP
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Pierce High Sensitivity NeutrAvidin HRP Conjugate is a specially prepared peroxidase-conjugated form of avidin biotin-binding protein that provides signal amplification like poly-HRP and exceptional storage stability.
Highlights:
NeutrAvidin Protein – a specially deglycosylated form of avidin that provides highly specific, low-background binding of biotinylated antibodies in many applications
High sensitivity – detect low levels of target without background; obtain high signal-to-noise ratios
Cost effective – use less conjugate in Western blotting and ELISA applications and still obtain excellent results
Flexible – compatible with typical chemiluminescent, fluorescent and colorimetric peroxidase substrates
Convenient – ready-to-use stabilized liquid format means there is no waiting for thawing and no need to aliquot
Specifications
Specifications
Quantity | 5 mL |
Target Molecule | Biotin |
Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2°C to 8°C. |
Specificity | Biotin |
Detection Method | Colorimetric, Chemiluminescence, Chemifluorescence |
Enzyme | HRP (Horseradish Peroxidase) |
Form | Liquid |
For Use With (Application) | Western Blotting, ELISA |
Product Line | Pierce™ |
Label or Dye | HRP (Horseradish Peroxidase) |