Learn More
Thermo Scientific™ Pierce™ Pyrogen-Free Plate Strips with Frame, 12 x 8 well
The endotoxin-free 12 x 8-well plates are supplied with one frame and contain <0.0005 EU/well endotoxin. The individually wrapped 8-well strips are ideal for assays of less than 96 tests at a time.
Brand:  Thermo Scientific™ A43888
Product Code. 17875187
  • 365.00 DKK / 10mL
Description
These endotoxin-free 12 x 8-well plates are supplied with one frame and contain <0.0005 EU/well endotoxin. The individually wrapped 8-well strips are ideal for assays of less than 96 tests at a time.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Strip Plate
Sterile
5 plates, 1 frame
Endotoxin level <0.0005 EU/well. 12 strip x 8 well 96-well plate, strips are individual wrapped.
Chromogenic LAL Endotoxin Assay, Kinetic Chromogenic LAL Assay and Kinetic Turbidimetric LAL Assay
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number0B30F01807 Certificate TypeCertificate of Analysis Date17/04/2024 Catalog NumberA43888
Lot Number0C91T52807 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog NumberA43888
Lot Number0DC1F82507 Certificate TypeCertificate of Analysis Date29/07/2023 Catalog NumberA43888
Lot NumberSPECS Certificate TypeCertificate of Analysis Date20/09/2022 Catalog NumberA43888
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Pierce™ Pyrogen-Free Plate Strips with Frame, 12 x 8 well > 10 mL/Vial