missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ TRITC (5/6-tetramethyl-rhodamine isothiocyanate), mixed isomer Product Code.: 11809370

Thermo Scientific™ TRITC (5/6-tetramethyl-rhodamine isothiocyanate), mixed isomer

Product Code. 11809370 Shop All Thermo Scientific Products
10 mg, 10mg
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 11809370

Brand: Thermo Scientific™ 46112

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Label antibodies, proteins and other molecules for fluorescence imaging (ex 546; em 572-615) using these activated derivatives of red-fluorescent rhodamine dye.

Thermo Scientific™ TRITC and Rhodamine Labeling Reagents and Kits are high-performance derivatives of rhodamine dye, activated for easy and reliable labeling of antibodies, proteins and other molecules for use as fluorescent probes.

Rhodamine dyes are used in fluorescence microscopy, flow cytometry and immunofluorescence-based assays such as Western blotting and ELISA. TRITC, NHS-rhodamine and Texas Red™ Sulfonyl Chloride are reactive towards primary amine groups on proteins, peptides and other biomolecules. Rhodamine dyes are often a mixture of isomers with reactive groups attached at the 5- and 6-positions of the bottom ring. The properties of these isomers are indistinguishable in terms of excitation and emission spectra and for protein applications there is no need to isolate a specific isomer. The single isomer is used in high resolution separation techniques such as sequencing and capillary electrophoresis.

Highlights:

  • Label and purify antibody in about one hour Efficiently labels antibodies and other purified proteins at primary amines (lysine side chains) Standard protocol results in antibodies with excellent dye:protein ratios for optimum activity and fluorescence Single-use fluors: no need to weigh tiny amounts powder; kits contain single-use vials of reagent Efficient purification and removal of non-reacted dye with excellent protein recovery

Includes:

NHS-Rhodamine Antibody Labeling Kit (Mfr. No. 53031)

NHS-Rhodamine, 3 vials; Borate Buffer, 1mL; Purification Resin, 5mL; Spin Columns, 6 each; Collection Tubes, 12 each

TRUSTED_SUSTAINABILITY

Specifications

Description Pierce TRITC
Quantity 10 mg
Product Type TRITC
Solubility DMF (Dimethylformamide)
Content And Storage Storage: Upon receipt store at -20°C. Product is shipped at ambient temperature. Store fluorescent dye in foil pouch with desiccant to protect from light and moisture.
Chemical Reactivity Amine
Reactive Moiety Isothiocyanate
Label or Dye Rhodamine Isomers, TMR (Tetramethylrhodamine), TRITC Isomers
Label Type Classic Dyes
Product Line Pierce™
Emission 575
Excitation 552
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberWD325684 Certificate TypeCertificate of Analysis Date06/04/2021 Product Code46112
Lot NumberWA320753 Certificate TypeCertificate of Analysis Date27/01/2021 Product Code46112
Lot NumberWA320637 Certificate TypeCertificate of Analysis Date08/01/2021 Product Code46112
Lot NumberSC248123 Certificate TypeCertificate of Analysis Date19/07/2017 Product Code46112
Lot NumberSC248123A Certificate TypeCertificate of Analysis Date19/07/2017 Product Code46112
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ TRITC (5/6-tetramethyl-rhodamine isothiocyanate), mixed isomer >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.