Learn More
Elma™ Pin Sets
Modular basket system for the Elmasonic P, Elmasonic Select, and EASY.
115.00 DKK - 384.00 DKK
Specifications
Quantity 10 Pins/Set
Type Pin Set
For Use With (Equipment) Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
View More Specs
Products 8
Product Code Brand Model Price Quantity & Availability  
Product Code Brand Model Price Quantity & Availability  
17881307
View Documents
Elma™
1125209
0509
115.00 DKK
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
17841367
View Documents
Elma™
1125210
0158
115.00 DKK
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
17811317
View Documents
Elma™
1125211
0767
115.00 DKK
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
17861367
View Documents
Elma™
1125212
0613/0614
384.00 DKK
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
17891347
View Documents
Elma™
1125213
0772
115.00 DKK
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
17801317
View Documents
Elma™
1125214
0741
115.00 DKK
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
17821357
View Documents
Elma™
1125215
0001R
115.00 DKK
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
17821317
View Documents
Elma™
1125216
0021R
115.00 DKK
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
Description
Description
  • Stainless steel with plastic handles (10 to 120 and 180 to 300) or Rilsan coating (150)
  • Ergonomic handles
  • Autoclavable
  • Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
  • Square openings in the side walls for mounting silicone holders
Specifications
Specifications
10 Pins/Set
Pin Set
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Elma™ Pin Sets