missing translation for 'onlineSavingsMsg'
Learn More
Piperidine, ≥99.5%, Honeywell™ Product Code.: 15685860

Piperidine, ≥99.5%, Honeywell™

Product Code. 15685860
450 mL, 450mL
Click to view available options
Quantity:
200 mL
450 mL
Unit Size:
200mL
450mL
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 15685860

Brand: Honeywell Burdick & Jackson 571261450ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Chemical Identifiers

CAS 110-89-4
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD00005979
InChI Key NQRYJNQNLNOLGT-UHFFFAOYSA-N
Synonym hexahydropyridine, azacyclohexane, cyclopentimine, cypentil, hexazane, pentamethyleneimine, piperidin, pentamethylenimine, perhydropyridine, pentamethyleneamine
PubChem CID 8082
ChEBI CHEBI:18049
IUPAC Name piperidine
SMILES C1CCNCC1

Specifications

Density 0.862g/mL at 20°C
Flash Point 16°C (60.8°F)
Quantity 450 mL
UN Number UN2401
Beilstein 102438
Formula Weight 85.15g/mol
Percent Purity ≥99.5%
Chemical Name or Material Piperidine
compliance-icons compliance-icons compliance-icons
Product Identifier
  • piperidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : piperidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Piperidine, ≥99.5%, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.