Learn More
Sartorius Pipet Stands and Accessories
Optimize the use of mLINE™ and Proline™ mechanical pipets. Sartorius Pipet Stands and Accessories include single- and multi-pipet stands, colored-coded caps and a calibration tool. Can also be used with a range of standard pipet brands.
Brand:  Sartorius 726001
 View more versions of this product
Product Code. 11614702
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
Pipette Stands
  • Stores pipettes in an upright position while not in use to avoid contamination from the desktop
    • Linear Stand is designed for all mechanical devices and stores them neatly in a line for instant access
    • Carousel stand is ideal for saving bench space and stores up to 6 mechanical pipettes
    • mLINE/Proline Plus Single Holder compactly holds a single mechanical pipette
Color Coded Caps
  • Colored caps help identify the volume of mLINE pipettes so it can easily be matched to the corresponding tip size
  • Includes 5 color-coded replacement caps
Calibration Tool
  • Calibration Tool for mLINE or Proline Plus mechanical pipettes for continuous accuracy and precision
  • Can also be used as a tube opener
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Color Coded Caps
Color coding mLINE pipets by volume
5/Pk.
Gray/Yellow/Blue/Green/Red
mLINE™/Proline™ Plus mechanical pipette