Learn More
BRAND™ Pipette Tray
Drawer organizer
Brand:  BRAND™ 28300
199.21 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10261721
  • 205.00 DKK / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Description
  • PVC
  • Four compartments for approximately 30 graduated pipettes (1-20 mL)
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    PVC
    280 mm
    Product Title
    BRAND™ Pipette Tray > Dimensions (L x W): 280 x 220 mm