Learn More
Plas Labs™ Broome-Style Rodent Restrainers
Ideal for injections or blood sampling
Brand:  Plas Labs™ 554-BSRR
 View more versions of this product
Product Code. 15193745
  • 1530.00 DKK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Offer easy access to rodent
  • Prevent rodent from turning
  • Provide safety to the animal handler
  • Manufactured from clear materials to provide complete visibility of animal
Specifications
Specifications
Clear
Rodent Restrainer
Chemically sterilized or washed at 180°F
8.5 in.
2.5 in.
250 to 500 g
Injections or Blood Sampling
Noryl™ Plastic
Broome
21.59 cm
6.35 cm