Learn More
Herenz Heinz™ Plastic Envelope Bag
Brand: Herenz Heinz™ 1110503
This item is not returnable.
View return policy
Specifications
Envelope Bag | |
Plastic | |
Zipper | |
300 x 210 x 30 mm |
Product Content Correction
Product Title
Herenz Heinz™ Plastic Envelope Bag > Size: 300 x 210 x 30mm
Spot an opportunity for improvement?Share a Content Correction