Learn More
Cole-Parmer™ Indenti-plug™ Plastic Foam Stopper
Keep the contents of your containers secure with these durable and reusable stoppers. Indenti-plug™ Plastic Foam Stoppers can also be used as a cleaner for pipettes, tubes, vials and precision tubing.
Brand:  Cole-Parmer™ 06298-80
 View more versions of this product
Product Code. 11735289
  • 955.00 DKK / Pack of 80
Estimated Shipment: 24-06-2025
to see stock.
Explore more special offers
Description
  • Breathable, nonabsorbent, chemical resistant and heat tolerant
  • Open-cell foam has a uniform density
  • Leaves no fibers
  • Autoclavable
Specifications
Specifications
45 to 50 mm
Autoclavable
Plastic
80/Pk.
White
Plug Stopper
Product Title
Cole-Parmer™ Indenti-plug™ Plastic Foam Stopper > Inner Diameter: 45 to 50mm