Learn More
DWK Life Sciences Plastic Hose Connection, with silicone seal (VMQ), PP complete with screw cap PBT, straight
For safe connection of hoses.
Brand:  DWK Life Sciences 286221806
2569.75 DKK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Product Code. 15804628
  • 2705.00 DKK / Pack of 100
In Stock
to see stock.
Explore more special offers
Description
Hoses can be securely connected to laboratory apparatus using plastic nozzles screwed onto a glass thread using a screw cap with bore. Plastic nozzles made of PP are used for lower thermal and chemical resistance requirements.
  • Hose olives enable the connection of hoses to laboratory apparatuses
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100/Pk.
8.6 mm
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    27
    Hours
    18
    Minutes
    29
    Seconds
    21
Product Content Correction
Product Title
DWK Life Sciences Plastic Hose Connection, with silicone seal (VMQ), PP complete with screw cap PBT, straight > GL 14