Learn More
Kartell™ Plastilab™ Untapered Y-Shaped Tubing Connectors
BIOPHARMA
Designed for use with hose system and vacuum lines.
Brand:  Kartell™ 0046600
Product Code. 12589656
  • 713.00 DKK / Pack of 100
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Designed for use with hose system and vacuum lines.
  • Made from polypropylene
  • Autoclavable
  • Nominal dimensions
Compliance
Meet Food and Agriculture regulations
Specifications
Specifications
Meet with the Food and Drug regulations. 90/128/CEE 92/39/CEE e FDA 21 CFR 177.1520, CE and IVD
Autoclavable
Useful for any hoses system and vacuum line, suitable for food stuff
4.6mm bore
Untapered Y Tubing Connector
Polypropylene
1
Y