Learn More
Grant Instruments™ Platform for Grant Orbital Shakers and Shaker Incubators
Includes clamps to keep flasks and beakers from spilling. Grant Instruments™ Platform for Grant Orbital Shakers and Shaker Incubators are interchangeable to accommodate a variety of vessels.
Brand:  Grant Instruments™ P9-500
3267.90 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11441068
  • 3355.00 DKK / Each
Only null left
Estimated Shipment: 24-09-2025
Description
  • Includes clamps to keep vessels secure
  • For use with
    • ES-20 Shaker Incubator
    • ES-80 Shaker Incubator
    • PSU 10i Orbital Shaker
    • PSU 20i Orbital Shaker
    TRUSTED_SUSTAINABILITY
Specifications
Specifications
9 x 500 mL Flasks
PSU-20i Orbital Platform Shaker
9
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Grant Instruments™ Platform for Grant Orbital Shakers and Shaker Incubators > For Use With: 9 x 500mL flasks