missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ă–GUSSA Platinum Crucible With Lid
Available in 10mL to 50mL capacities
Brand: Ă–GUSSA 8411 2030
This item is not returnable.
View return policy
Description
Quoted masses are approximate.Approximate process temperature 1,200°C and maximum temperature is 1,400°C (for short time of 10min)
Specifications
Platinum | |
1Each | |
35mm | |
Crucible |
1200°C | |
30 mL | |
40mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ă–GUSSA Platinum Crucible With Lid >
Spot an opportunity for improvement?Share a Content Correction