Learn More
Cole-Parmer Platinum-Cured Silicone Tubing, 61 Shore A
E in., hibits very good resilience, kink resistance, and pressure resistance
Brand:  Cole-Parmer 50119-57
 View more versions of this product
Product Code. 30053638
  • 1310.00 DKK / 15m
Only null left
Estimated Shipment: 04-08-2025
Add to basket
This item is not returnable. View return policy
Description
Description
Superior optical clarity—easily view your sample. Odorless, noncytoto in., ic, and nonhemolytic. Low-binding surface helps minimize particle and odor absorption. Does not contain BPA, phthalates, PVC, or pero in., ides. Cleanroom packed and double bagged package. Sterilize by autoclave, gamma irradiation, EtO, and chemical sterilants. Complies with U.S. Pharmacopeia 43, National Formulary 38 (USP), General Chapter 88 Class VI, Biological reactivity Tests, In Vivo (2020). Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Specifications
Specifications
ADCF;FDA;Non-Phthalate;USP;RoHS
3/16 in.
5/16 in.
50 ft.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer Platinum-Cured Silicone Tubing, 61 Shore A > 3/16 in., 5/16 in.