missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ II Taq Hot-Start DNA Polymerase

Invitrogen Platinum II Taq Hot-Start DNA Polymerase is designed for universal primer annealing and fast, easy PCR with its unique combination of innovative buffer, high-performance engineered Taq DNA polymerase, and superior hot-start technology.

Brand:  Invitrogen™ 14966001

 View more versions of this product

Product Code. 15826325

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Includes: 40 ÎĽL Platinum II Taq HS DNA Polymerase, 1.25 mL 5X Platinum II PCR Buffer, 1.25 mL Platinum GC Enhancer
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

• Platinum II Taq HS DNA Polymerase, 40 μL
• 5X Platinum II PCR Buffer, 1.25 mL
• Platinum GC Enhancer, 1.25 mL

Store at-20°C in a non-frost-free freezer.
High
Fast or Standard
100 reactions
Hot-start PCR
1X
100 Reactions
Separate Components
DNA
Tube
Platinum II Taq Hot-Start DNA Polymerase
Hot Start DNA Polymerase
Dry Ice
2X
Built-In Hot Start
3'-A
5 kb or less
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2805021 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number14966001
Lot Number2825340 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number14966001
Lot Number2796037 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number14966001
Lot Number2837319 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number14966001
Lot Number2837333 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number14966001
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Platinum™ II Taq Hot-Start DNA Polymerase > 100 Reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.