missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHA2 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-06069AF405
This item is not returnable.
View return policy
Description
PLEKHA2 Polyclonal specifically detects PLEKHA2 in Human samples. It is validated for Western Blot.Specifications
PLEKHA2 | |
Polyclonal | |
Western Blot | |
FLJ25921, PH domain-containing family A member 2, pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 2, pleckstrin homology domain-containing family A member 2, pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 2, tandem PH Domain containing protein-2, tandem PH domain-containing protein 2, TAPP2, TAPP-2 | |
Produced in rabbits immunized with E. coli-derived Human PLEKHA2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
59339 | |
Store at 4°C in the dark. | |
IgG |