missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Plug-In Tube Extension for SF110plus, Straight

Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.

Brand:  Memmert™ SF110plusB29718

Product Code. 17528095

  • 328.00 DKK / Each
Estimated Shipment: 14-10-2024
to see stock.
Add to basket
This item is not returnable. View return policy

Description

Description

  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight
Specifications

Specifications

60.3 mm
SF110plus Sterilizer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers