Learn More
Memmert™ Plug-In Tube Extension for SF750, Straight
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
Brand: Memmert™ SF750B29719
This item is not returnable.
View return policy
Description
- Accessory
- Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
- Straight
Specifications
60.3 mm | |
SF750 Sterilizer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Plug-In Tube Extension for SF750, Straight > For Use With: SF750
Spot an opportunity for improvement?Share a Content Correction