missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Plug-In Tube Extension for UF1060, Straight
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
Brand: Memmert™ UF1060B29719
This item is not returnable.
View return policy
Description
- Accessory
- Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
- Straight
Specifications
UF1060 Oven | |
Plug-in Tube Extension | |
O.D. 60.3 mm, I.D. 57 mm |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Plug-In Tube Extension for UF1060, Straight > For Use With: UF1060
Spot an opportunity for improvement?Share a Content Correction