missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Plug-In Tube Extension for UF1060, Straight
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
Brand: Memmert™ UF1060B29718
This item is not returnable.
View return policy
Description
- Accessory
- Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
- Straight
Specifications
UF1060 Oven | |
Plug-in Tube Extension | |
O.D. 60.3 mm, I.D. 57 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Plug-In Tube Extension for UF1060, Straight > For Use With: UF1060
Spot an opportunity for improvement?Share a Content Correction