Learn More
Memmert™ Plug-In Tube Extension for UF260, Straight
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
Brand:  Memmert™ UF260B29718
Product Code. 17529115
  • 354.00 DKK / Each
Estimated Shipment: 03-04-2025
to see stock.
Explore more special offers
Description
  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight
Specifications
Specifications
UF260 Oven
Plug-in Tube Extension
O.D. 60.3 mm, I.D. 57 mm
Product Title
Memmert™ Plug-In Tube Extension for UF260, Straight > For Use With: UF260