Learn More
Molecular Probes™ Pluronic™ F-127, 0.2 μm filtered (10% Solution in Water)
Pluronic™ F-127, 0.2 μm filtered (10% Solution in Water)
Brand: Molecular Probes™ P6866
1403.00 DKK valid until 2025-08-31
Use promo code "24157" to get your promotional price.
Description
Pluronic F-127 *10% solution in water* is a nonionic, surfactant polyol (molecular weight approximately 12,500 daltons) that has been found to facilitate the solubilization of water-insoluble dyes and other materials in physiological media. Pluronic F-127 is commonly used to help disperse the acetoxymethyl (AM) esters of our ion indicators as well as our cell tracer dyes such as CFDA-SE.
Specifications
Colorless | |
Liquid | |
30 mL |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Promotions
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24157Expires: 2025-08-31
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.