Description
Kit includes the PNGase F enzyme, which cleaves N-glycan chains at the innermost GlcNAc and asparagine residues of high mannose, hybrid, and complex oligosaccharides.
Features:
- ≥95% purity
- Recombinant PNGase F enzyme with no endoglycosidase F1, F2, or F3
- Enzyme leaves N-glycan core oligosaccharides intact and suitable for downstream analysis
- Suitable for use under native or denaturing conditions
Components included in the PNGase F Glycan Cleavage Kit are: 33,000 units of PNGase F (500,000 units/mL) and 1 mL PNGase F 10X buffer
Specifications
Specifications
Content And Storage | • 30,000 units PNGase F enzyme, recombinant (500,000 units/mL), store at -20°C • 1 mL PNGase F 10X Buffer, store at -20°C |
Shipping Condition | Wet Ice |
Form | Liquid |
Quantity | 1 kit |
Product Type | PNGase F Glycan Cleavage Kit |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Gibco™ PNGase F Glycan Cleavage Kit >
Spot an opportunity for improvement?Share a Content Correction