Learn More
Bellingham Stanley™ Polarimeter Tube
Brand:  Bellingham Stanley™ 35-30
2089.07 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11994219
  • 2530.00 DKK / Each
Only null left
Estimated Shipment: 25-08-2025
Specifications
Polarimeter Tube
Glass
Air Bubble
200 mm
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Bellingham Stanley™ Polarimeter Tube > Length: 200mm