Learn More
Buerkle™ Polyacetal Hose Clips
Use these Buerkle™ Polyacetal Hose Clips to clamp down on tubing for a sure and secure fit.
Brand: Buerkle™ 8678-0012
This item is not returnable.
View return policy
Description
- Hose clips made of Polyacetal (POM)
- Featured in varying diameters
- POM is white, sterilizable, does not rust, permanently elastic and hygienic.
Specifications
Polyacetal | |
10/Pk.; 10/Scale | |
Hose Clip |
Hoses | |
11 to 12 mm |
Product Content Correction
Product Title
Buerkle™ Polyacetal Hose Clips > For Diameter: 11 to 12mm
Spot an opportunity for improvement?Share a Content Correction