Learn More
HUBER Polycarbonate Bath
A range of polycarbonate baths in various capacities
Brand: HUBER 30528
2872.02 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Polycarbonate bathSpecifications
Bath | |
Polycarbonate | |
108 A |
Product Content Correction
Product Title
HUBER Polycarbonate Bath > Model: 108A
Spot an opportunity for improvement?Share a Content Correction