Learn More
HUBER Polycarbonate Bath
A range of polycarbonate baths in various capacities
Brand:  HUBER 17098
6185.88 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10374245
  • 7145.00 DKK / Each
Only null left
Estimated Shipment: 27-08-2025
Description
Polycarbonate bath
Specifications
Specifications
Bath
Polycarbonate
130 A
Certificates
Promotions
Promotions
Product Content Correction
Product Title
HUBER Polycarbonate Bath > Model: 130A