Learn More
Bryson Packaging™ Polyethene Bags
Resealable bags with write-on patch for labelling
55.97 DKK - 773.00 DKK
Specifications
Product Type | Bag |
---|---|
Closure Type | Grip Seal |
Color | Clear |
For Use With (Application) | Storing stationery |
Material | Polyethylene |
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | ||||||
30057396
|
Bryson Packaging™
BAJ-360-110V |
75 mm |
773.00 DKK
Pack of 1000 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
11372274
|
Bryson Packaging™
BAJ-360-110V |
75 mm | N/A | N/A | N/A | |||||
30057393
|
Bryson Packaging™
BAJ-360-150J |
50 mm |
617.00 DKK
Pack of 1000 |
Estimated Shipment: 02-07-2025
Log in to see stock. |
||||||
11312284
|
Bryson Packaging™
BAJ-360-150J |
50 mm | N/A | N/A | N/A | |||||
Description
Polyethylene, resealable bags, clear or with three write-on bars for labelling. Ideal for storing stationery and sending samples etc.Specifications
Bag | |
Clear | |
Polyethylene | |
100 Pack |
Grip Seal | |
Storing stationery | |
100 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Bryson Packaging™ Polyethene Bags