missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene glycol 400, EssentQ, Scharlab
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Specifications
Specifications
CAS | 25322-68-3 |
Molecular Formula | (C2H4O)n |
MDL Number | MFCD01779601 |
Quantity | 1 L |
InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
SMILES | [H]OCCO |
IUPAC Name | ethane-1,2-diol |
Molecular Weight (g/mol) | 62.07 |
Grade | EssentQ |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Polyethylene glycol 400, EssentQ, Scharlab >
Spot an opportunity for improvement?Share a Content Correction