missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol 600 Grade, Extra Pure, SLR, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
Packaging:
Amber glass bottle
Unit Size:
500mL
Specifications
Specifications
pH | 5.0 to 7.0 |
CAS | 25322-68-3 |
Physical Form | Liquid |
Melting Point | 17°C |
Molecular Formula | (C2H4O)n |
MDL Number | MFCD01779601 |
Quantity | 500 mL |
InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
SMILES | [H]OCCO |
IUPAC Name | ethane-1,2-diol |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Polyethylene Glycol 600 Grade, Extra Pure, SLR, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction