Learn More
Kautex™ Polyethylene Wide Mouth Square Bottles
Manufactured with HDPE for strength and durability. Kautex™ Polyethylene Wide Mouth Square Bottles are ideal for packaging, storage and transporting media.
Brand: Kautex™ 2000070539
This item is not returnable.
View return policy
Description
- Easy filling, emptying and cleaning
- Ribbed sides for a secure grip
- Rectangular labelling field
- Rectangular shape optimizes space utilisation
- Leakproof
Specifications
500 mL | |
30.8 mm | |
HDPE | |
164/Cs. | |
145.5 mm |
Natural | |
Packaging, Storage, Transport | |
164 | |
Square | |
Wide Neck Bottle |
Product Content Correction
Product Title
Kautex™ Polyethylene Wide Mouth Square Bottles > Capacity: 500 mL; Height: 145.5 mm; Color: Natural
Spot an opportunity for improvement?Share a Content Correction